α-Cyclopropyl-4-ethylbenzenemethanol ≥96%

Supplier: AOBChem USA

259295-1G 259295-500MG 259295-5G 259295-25G
77467-624EA 341.96 USD
77467-624 77480-332 77490-660 77500-782
α-Cyclopropyl-4-ethylbenzenemethanol ≥96%
α-Cyclopropyl-4-ethylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆O
MDL Number: MFCD00013732
CAS Number: 84100-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR