α-Cyclopropyl-4-(1,1-dimethylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

259293-1G 259293-500MG 259293-5G 259293-25G
77467-620EA 397.58 USD
77467-620 77480-328 77490-656 77500-778
α-Cyclopropyl-4-(1,1-dimethylethyl)benzenemethanol ≥95%
α-Cyclopropyl-4-(1,1-dimethylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₀O
MDL Number: MFCD00114260
CAS Number: 83763-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR