α-Cyclopropyl-4-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

259286-1G 259286-500MG 259286-5G 259286-25G
77467-616EA 371.83 USD
77467-616 77480-324 77490-652 77500-774
α-Cyclopropyl-4-methylbenzenemethanol ≥95%
α-Cyclopropyl-4-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄O
MDL Number: MFCD00019254
CAS Number: 6552-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR