4-Fluoro-α,α-dipropylbenzenemethanol ≥97%

Supplier: AOBChem USA

259209-1G 259209-500MG 259209-5G 259209-25G
77467-612EA 397.58 USD
77467-612 77480-320 77490-648 77500-770
4-Fluoro-α,α-dipropylbenzenemethanol ≥97%
4-Fluoro-α,α-dipropylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₉FO
MDL Number: MFCD34875710
CAS Number: 76499-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR