3-Bromo-2,6-difluoro-α-(1-methylbutyl)benzenemethanol ≥95%

Supplier: AOBChem USA

259061-1G 259061-500MG 259061-5G 259061-25G
77467-580EA 415.09 USD
77467-580 77480-288 77490-616 77500-746
3-Bromo-2,6-difluoro-α-(1-methylbutyl)benzenemethanol ≥95%
3-Bromo-2,6-difluoro-α-(1-methylbutyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₅BrF₂O
MDL Number: MFCD34875686
CAS Number: 1692071-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR