3,5-Dimethyl-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

25804-1G 25804-500MG 25804-5G 25804-25G
77467-552EA 368.74 USD
77467-552 77480-262 77490-586 77500-720
3,5-Dimethyl-2-methoxyphenylboronic acid ≥97%
3,5-Dimethyl-2-methoxyphenylboronic acid

Precaución: For research use only

Formula: C₉H₁₃BO₃
MDL Number: MFCD22414932
CAS Number: 1451391-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR