6-Bromo-3-methyl-2-nitrophenol ≥96%

Supplier: AOBChem USA

257112-1G 257112-500MG 257112-5G 257112-25G
77467-540EA 248.23 USD
77467-540 77480-250 77490-574 77500-710
6-Bromo-3-methyl-2-nitrophenol ≥96%
6-Bromo-3-methyl-2-nitrophenol

Precaución: For research use only

Formula: C₇H₆BrNO₃
MDL Number: MFCD28745602
CAS Number: 1195533-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR