propyl 3-hydroxypentanoate ≥95%

Supplier: AOBChem USA

255784-1G 255784-500MG 255784-5G
77467-512EA 589.16 USD
77467-512 77480-224 77490-546
propyl 3-hydroxypentanoate ≥95%
propyl 3-hydroxypentanoate

Precaución: For research use only

Formula: C₈H₁₆O₃
MDL Number: MFCD34874512
CAS Number: 133098-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR