4-Methylbiphenyl-3-ylboronic acid ≥97%

Supplier: AOBChem USA

25508-1G 25508-500MG 25508-5G 25508-25G
77467-508EA 339.9 USD
77467-508 77480-220 77490-542 77500-686
4-Methylbiphenyl-3-ylboronic acid ≥97%
4-Methylbiphenyl-3-ylboronic acid

Precaución: For research use only

Formula: C₁₃H₁₃BO₂
MDL Number: MFCD23713126
CAS Number: 1438810-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR