2,6-Difluoro-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

25495-1G 25495-500MG 25495-5G 25495-25G
77467-504EA 195.7 USD
77467-504 77480-216 77490-538 77500-682
2,6-Difluoro-3-(methylthio)benzoic acid ≥97%
2,6-Difluoro-3-(methylthio)benzoic acid

Precaución: For research use only

Formula: C₈H₆F₂O₂S
MDL Number: MFCD22490214
CAS Number: 1428234-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR