5-Chloro-2-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

25281-5G 25281-25G
77490-530EA 512.94 USD
77490-530 77500-674
5-Chloro-2-phenylpyridine ≥97%
5-Chloro-2-phenylpyridine

Precaución: For research use only

Formula: C₁₁H₈ClN
Storage Temperature: Ambient
MDL Number: MFCD04114185
CAS Number: 58254-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR