3-Bromo-5-phenylsalicylic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
252712-5G
252712-25G
77490-526EA
1454.36
USD
77490-526
77500-670
3-Bromo-5-phenylsalicylic acid ≥95%
3-Bromo-5-phenylsalicylic acid
Precaución: For research use only
Formula:
C₁₃H₉BrO₃ Storage Temperature: Ambient |
MDL Number:
MFCD18378258 CAS Number: 4906-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...