2,4-Dibromo-6-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

25161-1G 25161-500MG 25161-5G
77467-484EA 457.32 USD
77467-484 77480-200 77490-518
2,4-Dibromo-6-(trifluoromethyl)benzaldehyde ≥95%
2,4-Dibromo-6-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₈H₃Br₂F₃O
MW: 331.912 g/mol
MDL Number: MFCD34755809
CAS Number: 2167236-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR