3-Fluoro-5-trifluoromethylthioanisole ≥97%

Supplier: AOBChem USA

24861-1G 24861-500MG 24861-5G 24861-25G
77467-464EA 226.6 USD
77467-464 77480-182 77490-494 77500-642
3-Fluoro-5-trifluoromethylthioanisole ≥97%
3-Fluoro-5-trifluoromethylthioanisole

Precaución: For research use only

Formula: C₈H₆F₄S
MDL Number: MFCD22628231
CAS Number: 1414870-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR