4-(Cyclopropylmethoxy)-2-methylbenzoic acid ≥96%

Supplier: AOBChem USA

248222-1G 248222-500MG 248222-5G
77467-456EA 452.17 USD
77467-456 77480-174 77490-486
4-(Cyclopropylmethoxy)-2-methylbenzoic acid ≥96%
4-(Cyclopropylmethoxy)-2-methylbenzoic acid

Precaución: For research use only

Formula: C₁₂H₁₄O₃
MDL Number: MFCD24079451
CAS Number: 1224719-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR