2,4,6-Trimethyl-α-(2,4,6-trimethylphenyl)benzenemethanol ≥95%

Supplier: AOBChem USA

247115-1G 247115-500MG 247115-5G 247115-25G
77467-432EA 156.56 USD
77467-432 77480-152 77490-462 77500-614
2,4,6-Trimethyl-α-(2,4,6-trimethylphenyl)benzenemethanol ≥95%
2,4,6-Trimethyl-α-(2,4,6-trimethylphenyl)benzenemethanol

Precaución: For research use only

Formula: C₁₉H₂₄O
MDL Number: MFCD01463301
CAS Number: 21127-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR