2,3,6-Trimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24699-5G 24699-25G
77490-454EA 811.64 USD
77490-454 77500-606
2,3,6-Trimethoxybenzaldehyde ≥97%
2,3,6-Trimethoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD06739499
CAS Number: 5556-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR