3,4'-Dimethylbenzhydrol ≥96%

Supplier: AOBChem USA

Warning

Synonyms: 3-Methyl-α-(4-methylphenyl)benzenemethanol

246744-5G 246744-25G
77490-434EA 1082.53 USD
77490-434 77500-592
3,4'-Dimethylbenzhydrol ≥96%
3,4'-Dimethylbenzhydrol

Precaución: For research use only

Formula: C₁₅H₁₆O
MW: 212.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201230
CAS Number: 13389-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR