3-Fluoro-4'-methoxybenzhydrol ≥96%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-α-(4-methoxyphenyl)benzenemethanol

246716-5G 246716-25G
77490-430EA 334.75 USD
77490-430 77500-588
3-Fluoro-4'-methoxybenzhydrol ≥96%
3-Fluoro-4'-methoxybenzhydrol

Precaución: For research use only

Formula: C₁₄H₁₃FO₂
MW: 232.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201328
CAS Number: 38158-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR