Methanone, cyclopropyl(2,3-dimethylphenyl)- ≥95%

Supplier: AOBChem USA

246503-1G 246503-500MG 246503-5G 246503-25G
77467-388EA 382.13 USD
77467-388 77480-108 77490-418 77500-578
Methanone, cyclopropyl(2,3-dimethylphenyl)- ≥95%
Methanone, cyclopropyl(2,3-dimethylphenyl)-

Precaución: For research use only

Formula: C₁₂H₁₄O
MDL Number: MFCD03841221
CAS Number: 898790-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR