6'-Bromo-3'-chloro-2'-fluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

246496-5G 246496-25G
77490-414EA 587.1 USD
77490-414 77500-574
6'-Bromo-3'-chloro-2'-fluoroacetophenone ≥95%
6'-Bromo-3'-chloro-2'-fluoroacetophenone

Precaución: For research use only

Formula: C₈H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD28096615
CAS Number: 1540199-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR