1-(4-Chloro-2-fluorophenyl)-1-propanone ≥96%

Supplier: AOBChem USA

246448-1G 246448-500MG 246448-5G 246448-25G
77467-380EA 82.4 USD
77467-380 77480-100 77490-410 77500-570
1-(4-Chloro-2-fluorophenyl)-1-propanone ≥96%
1-(4-Chloro-2-fluorophenyl)-1-propanone

Precaución: For research use only

Formula: C₉H₈ClFO
MDL Number: MFCD13194252
CAS Number: 186685-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR