5-Bromo-α-cyclopropyl-2-fluoro-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

246235-1G 246235-500MG 246235-5G 246235-25G
77467-376EA 312.09 USD
77467-376 77480-096 77490-406 77500-564
5-Bromo-α-cyclopropyl-2-fluoro-α-methylbenzenemethanol ≥97%
5-Bromo-α-cyclopropyl-2-fluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₂BrFO
MDL Number: MFCD23759092
CAS Number: 1508626-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR