2,6-Dimethoxypyridine-3-methanol ≥97%

Supplier: AOBChem USA

24481-1G 24481-5G 24481-25G
77467-344EA 62.83 USD
77467-344 77490-374 77500-532
2,6-Dimethoxypyridine-3-methanol ≥97%
2,6-Dimethoxypyridine-3-methanol

Precaución: For research use only

Formula: C₈H₁₁NO₃
MDL Number: MFCD08276849
CAS Number: 562840-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR