3-Bromo-2-chloro-5-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

24463-5G 24463-25G
77490-370EA 877.56 USD
77490-370 77500-528
3-Bromo-2-chloro-5-methoxypyridine ≥95%
3-Bromo-2-chloro-5-methoxypyridine

Precaución: For research use only

Formula: C₆H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD10699708
CAS Number: 1033202-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR