3,5-Diethyltoluene ≥95%

Supplier: AOBChem USA

243643-5G 243643-25G
77490-362EA 1409.04 USD
77490-362 77500-522
3,5-Diethyltoluene ≥95%
3,5-Diethyltoluene

Precaución: For research use only

Formula: C₁₁H₁₆
CAS Number: 2050-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR