Ethyl-1-(cyanomethyl)-1H-pyrazole-3-carboxylate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 1-(cyanomethyl)-1H-pyrazole-3-carboxylate

243562-1G 243562-500MG 243562-5G 243562-25G
77467-328EA 245.14 USD
77467-328 77480-048 77490-356 77500-516
Ethyl-1-(cyanomethyl)-1H-pyrazole-3-carboxylate ≥95%
Ethyl-1-(cyanomethyl)-1H-pyrazole-3-carboxylate

Precaución: For research use only

Formula: C₈H₉N₃O₂
MW: 179.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15146498
CAS Number: 1217863-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR