1-(5-Bromo-2-fluorophenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

243496-1G 243496-500MG 243496-5G 243496-25G
77467-324EA 226.6 USD
77467-324 77480-046 77490-354 77500-514
1-(5-Bromo-2-fluorophenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(5-Bromo-2-fluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
MW: 261.1306 g/mol
MDL Number: MFCD21328331
CAS Number: 1488272-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR