4-Methyl-1-nitro-2-phenoxybenzene ≥97%

Supplier: AOBChem USA

243263-1G 243263-500MG 243263-5G 243263-25G
77467-320EA 154.5 USD
77467-320 77480-040 77490-348 77500-510
4-Methyl-1-nitro-2-phenoxybenzene ≥97%
4-Methyl-1-nitro-2-phenoxybenzene

Precaución: For research use only

Formula: C₁₃H₁₁NO₃
MDL Number: MFCD20133222
CAS Number: 60287-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR