3-(2,5-Difluorophenyl)-3-hydroxypropanenitrile ≥96%

Supplier: AOBChem USA

242962-1G 242962-500MG 242962-5G
77467-304EA 644.78 USD
77467-304 77480-028 77490-336
3-(2,5-Difluorophenyl)-3-hydroxypropanenitrile ≥96%
3-(2,5-Difluorophenyl)-3-hydroxypropanenitrile

Precaución: For research use only

Formula: C₉H₇F₂NO
MDL Number: MFCD21801237
CAS Number: 1505577-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR