α-Heptyl-4-methoxy-3,5-dimethylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

242229-1G 242229-5G 242229-25G
77467-292EA 415.09 USD
77467-292 77490-324 77500-496
α-Heptyl-4-methoxy-3,5-dimethylbenzenemethanol ≥96%
α-Heptyl-4-methoxy-3,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₇H₂₈O₂
Storage Temperature: Ambient
MDL Number: MFCD25866137
CAS Number: 1548879-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR