4-(Ethylthio)-α-heptylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

242162-1G 242162-500MG 242162-5G 242162-25G
77467-284EA 415.09 USD
77467-284 77480-008 77490-316 77500-488
4-(Ethylthio)-α-heptylbenzenemethanol ≥95%
4-(Ethylthio)-α-heptylbenzenemethanol

Precaución: For research use only

Formula: C₁₆H₂₆OS
MW: 266.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870649
CAS Number: 1976436-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR