4-Fluoro-α-heptylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

242148-1G 242148-500MG 242148-5G 242148-25G
77467-280EA 226.6 USD
77467-280 77480-004 77490-312 77500-484
4-Fluoro-α-heptylbenzenemethanol ≥97%
4-Fluoro-α-heptylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₁FO
MW: 224.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786854
CAS Number: 153644-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR