2,6-Difluoro-3-methyl-α-pentylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

242009-1G 242009-500MG 242009-5G 242009-25G
77467-272EA 415.09 USD
77467-272 77479-996 77490-304 77500-476
2,6-Difluoro-3-methyl-α-pentylbenzenemethanol ≥95%
2,6-Difluoro-3-methyl-α-pentylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈F₂O
MW: 228.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24310368
CAS Number: 1554846-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR