α-Pentyl[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

241918-1G 241918-500MG 241918-5G
77467-264EA 487.19 USD
77467-264 77479-988 77490-296
α-Pentyl[1,1'-biphenyl]-4-methanol ≥95%
α-Pentyl[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₁₈H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD18958998
CAS Number: 50673-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR