4-Bromo-α-pentylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

241864-1G 241864-500MG 241864-5G 241864-25G
77467-256EA 397.58 USD
77467-256 77479-980 77490-288 77500-464
4-Bromo-α-pentylbenzenemethanol ≥95%
4-Bromo-α-pentylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD12786822
CAS Number: 71434-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR