2-Fluoro-α-hexyl-5-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

241846-1G 241846-500MG 241846-5G 241846-25G
77467-252EA 415.09 USD
77467-252 77479-976 77490-284 77500-460
2-Fluoro-α-hexyl-5-methylbenzenemethanol ≥97%
2-Fluoro-α-hexyl-5-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₁FO
Storage Temperature: Ambient
MDL Number: MFCD19602387
CAS Number: 1343506-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR