α-Hexyl-3-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

241742-1G 241742-500MG 241742-5G 241742-25G
77467-244EA 415.09 USD
77467-244 77479-968 77490-274 77500-450
α-Hexyl-3-(methylthio)benzenemethanol ≥95%
α-Hexyl-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂OS
MW: 238.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870501
CAS Number: 2141543-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR