2-Bromo-3-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

24015-1G 24015-500MG 24015-5G 24015-25G
77467-236EA 324.45 USD
77467-236 77479-960 77490-266 77500-444
2-Bromo-3-(methylthio)pyridine ≥95%
2-Bromo-3-(methylthio)pyridine

Precaución: For research use only

Formula: C₆H₆BrNS
MW: 204.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702661
CAS Number: 884863-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR