5-Bromo-2-(2-chlorophenoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

23990-1G 23990-500MG 23990-5G 23990-25G
77467-232EA 253.38 USD
77467-232 77479-956 77490-262 77500-440
5-Bromo-2-(2-chlorophenoxy)pyridine ≥97%
5-Bromo-2-(2-chlorophenoxy)pyridine

Precaución: For research use only

Formula: C₁₁H₇BrClNO
MW: 284.536 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14652051
CAS Number: 1240670-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR