2-(4-Fluorophenyl)-2-methyl-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

23916-1G 23916-500MG 23916-5G 23916-25G
77467-228EA 81.37 USD
77467-228 77479-952 77490-258 77500-436
2-(4-Fluorophenyl)-2-methyl-1,3-dioxolane ≥95%
2-(4-Fluorophenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD19347731
CAS Number: 36881-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR