2-Chloro-6-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

23790-1G 23790-500MG 23790-5G 23790-25G
77467-224EA 282.22 USD
77467-224 77479-948 77490-254 77500-432
2-Chloro-6-phenylpyridine ≥97%
2-Chloro-6-phenylpyridine

Precaución: For research use only

Formula: C₁₁H₈ClN
MW: 189.641 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01646086
CAS Number: 13382-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR