1-Fluoro-4-methoxy-2-methylsulfanylbenzene ≥97%

Supplier: AOBChem USA

Warning

23725-1G 23725-500MG 23725-5G 23725-25G
77467-220EA 82.4 USD
77467-220 77479-944 77490-250 77500-430
1-Fluoro-4-methoxy-2-methylsulfanylbenzene ≥97%
1-Fluoro-4-methoxy-2-methylsulfanylbenzene

Precaución: For research use only

Formula: C₈H₉FOS
MW: 172.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489296
CAS Number: 836678-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR