2-(Methylthio)thiophene-4-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

23721-1G 23721-500MG 23721-5G 23721-25G
77467-216EA 282.22 USD
77467-216 77479-940 77490-246 77500-426
2-(Methylthio)thiophene-4-carboxaldehyde ≥95%
2-(Methylthio)thiophene-4-carboxaldehyde

Precaución: For research use only

Formula: C₆H₆OS₂
Storage Temperature: Ambient
MDL Number: MFCD22543668
CAS Number: 88511-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR