2,6-Dimethoxy-4-pyridinemethanol ≥97%

Supplier: AOBChem USA

Warning

23692-1G 23692-500MG 23692-5G 23692-25G
77467-212EA 258.53 USD
77467-212 77479-936 77490-242 77500-422
2,6-Dimethoxy-4-pyridinemethanol ≥97%
2,6-Dimethoxy-4-pyridinemethanol

Precaución: For research use only

Formula: C₈H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD18803411
CAS Number: 52606-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR