4-Methyl-2-(methylthio)thiophene ≥97%

Supplier: AOBChem USA

Warning

23657-1G 23657-500MG 23657-5G 23657-25G
77467-208EA 82.4 USD
77467-208 77479-932 77490-238 77500-418
4-Methyl-2-(methylthio)thiophene ≥97%
4-Methyl-2-(methylthio)thiophene

Precaución: For research use only

Formula: C₆H₈S₂
MW: 144.258 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22056777
CAS Number: 146041-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR