1-(3-Bromo-4-methoxy-benzyl)-piperidine ≥97%

Supplier: AOBChem USA

Warning

23591-1G 23591-500MG 23591-5G 23591-25G
77467-204EA 247.2 USD
77467-204 77479-928 77490-234 77500-412
1-(3-Bromo-4-methoxy-benzyl)-piperidine ≥97%
1-(3-Bromo-4-methoxy-benzyl)-piperidine

Precaución: For research use only

Formula: C₁₃H₁₈BrNO
MW: 284.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14645210
CAS Number: 886-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR