α-Ethenyl-2-fluoro-5-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235183-1G 235183-500MG 235183-5G
77467-200EA 466.59 USD
77467-200 77479-924 77490-230
α-Ethenyl-2-fluoro-5-methylbenzenemethanol ≥95%
α-Ethenyl-2-fluoro-5-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD34870255
CAS Number: 1603204-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR