5-Bromo-α-ethenyl-2-fluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

235149-1G 235149-500MG 235149-5G
77467-196EA 751.9 USD
77467-196 77479-920 77490-226
5-Bromo-α-ethenyl-2-fluorobenzenemethanol ≥97%
5-Bromo-α-ethenyl-2-fluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₈BrFO
MW: 231.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21232126
CAS Number: 149946-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR