α-Ethenyl-2,4-dimethoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235130-1G 235130-500MG 235130-5G 235130-25G
77467-192EA 380.07 USD
77467-192 77479-916 77490-222 77500-404
α-Ethenyl-2,4-dimethoxybenzenemethanol ≥95%
α-Ethenyl-2,4-dimethoxybenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21232084
CAS Number: 846563-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR